Assay Details
Target Gene Details
Entrez Gene ID: | 114088 |
Gene Name: | tripartite motif containing 9 |
Gene Aliases: |
RNF91, SPRING |
Location: |
Chr.14:50975261-51105173 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM9 | NM_015163.5 | 10 | 4449 | NP_055978.4 |
| XM_005267301.3 | 14 | 5255 | XP_005267358.1 | |
| XM_005267302.3 | 13 | 5066 | XP_005267359.1 | |
| XM_006720018.3 | 14 | 5309 | XP_006720081.1 | |
| XM_017020943.1 | 14 | 5297 | XP_016876432.1 | |
| XM_017020944.1 | 13 | 5120 | XP_016876433.1 | |
| XM_017020945.1 | 11 | 5054 | XP_016876434.1 | |
| XM_017020946.1 | 11 | 5042 | XP_016876435.1 | |
| XM_017020947.1 | 11 | 5000 | XP_016876436.1 | |
| XM_017020948.1 | 11 | 4988 | XP_016876437.1 | |
| XM_017020949.1 | 9 | 4757 | XP_016876438.1 | |
| XM_017020950.1 | 9 | 4703 | XP_016876439.1 | |
| BC063872.1 | 10 | 4806 | AAH63872.1 | |
| D87458.2 | 10 | 3826 | BAA13398.2 |
More Information
Set Membership: |
|

Genomic Map