Assay Details
Target Gene Details
Entrez Gene ID: | 151194 |
Gene Name: | methyltransferase like 21A |
Gene Aliases: |
FAM119A, HCA557b, HSPA-KMT |
Location: |
Chr.2:207580631-207625928 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3131 | Chr.2:207578177 - 207624543 on Build GRCh38 | Deletion |
|
| nsv1013908 | Chr.2:207577371 - 207629609 on Build GRCh38 | Loss |
|
| esv996264 | Chr.2:207608982 - 207616843 on Build GRCh38 | Deletion |
|
| esv2523513 | Chr.2:207609395 - 207612996 on Build GRCh38 | Deletion |
|
| esv3594090 | Chr.2:207608668 - 207651398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map