Assay Details
Target Gene Details
Entrez Gene ID: | 23603 |
Gene Name: | coronin 1C |
Gene Aliases: |
HCRNN4 |
Location: |
Chr.12:108645109-108731550 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO1C | NM_001105237.2 | 11 | 2012 | NP_001098707.1 |
| NM_001276471.1 | 11 | 2030 | NP_001263400.1 | |
| NM_014325.3 | 11 | 1931 | NP_055140.1 | |
| XM_011538125.1 | 7 | 1255 | XP_011536427.1 | |
| XM_017019121.1 | 12 | 2760 | XP_016874610.1 | |
| XM_017019122.1 | 11 | 2337 | XP_016874611.1 | |
| XM_017019123.1 | 12 | 2390 | XP_016874612.1 | |
| AB030656.1 | 11 | 1900 | BAA83077.1 | |
| AB209909.1 | 11 | 1886 | BAD93146.1 | |
| AK023205.1 | 11 | 1901 | ||
| AK096363.1 | 11 | 2030 | ||
| AK297463.1 | 11 | 1764 | ||
| AK311315.1 | ||||
| AK316065.1 | 11 | 1727 | ||
| AL162070.1 | 11 | 1869 | CAB82406.1 | |
| BC002342.2 | 11 | 1885 | AAH02342.1 |
More Information
Set Membership: |
|

Genomic Map