Assay Details
Target Gene Details
Entrez Gene ID: | 9703 |
Gene Name: | KIAA0100 |
Gene Aliases: |
BCOX, BCOX1, CT101, FMP27 |
Location: |
Chr.17:28614440-28645159 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0100 | NM_014680.3 | 35 | 6192 | NP_055495.2 |
| XM_005258073.2 | 35 | 6184 | XP_005258130.1 | |
| XM_017025423.1 | 34 | 6041 | XP_016880912.1 | |
| BC008591.2 | 5 | 496 | AAH08591.2 | |
| BC103726.1 | 5 | 618 | AAI03727.1 | |
| BC120873.1 | 35 | 6051 | ||
| CR933634.1 | 36 | 6051 | CAI45940.1 | |
| D43947.3 | 35 | 6164 | BAA07891.3 |
Target Gene Details
Entrez Gene ID: | 100506436 |
Gene Name: | SPAG5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:28599133-28617377 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG5-AS1 | NR_040012.1 | 3 | 674 | |
| AK098638.1 | 3 | 862 | ||
| AL157421.1 | 1 | 1003 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|

Genomic Map