Assay Details
Target Gene Details
Entrez Gene ID: | 11016 |
Gene Name: | activating transcription factor 7 |
Gene Aliases: |
ATFA |
Location: |
Chr.12:53507856-53626415 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7 | NM_001130060.1 | 12 | 2540 | NP_001123532.1 |
| NM_006856.2 | 12 | 2603 | NP_006847.1 | |
| NR_073163.1 | ||||
| XM_005268587.3 | 12 | 2555 | XP_005268644.1 | |
| XM_017018722.1 | 12 | 3743 | XP_016874211.1 | |
| AK298853.1 | ||||
| BP279417.1 | 1 | 268 | ||
| BP312300.1 | 1 | 350 | ||
| DB047160.1 | ||||
| X52943.1 | 12 | 2548 | CAA37118.1 |
Target Gene Details
Entrez Gene ID: | 100652999 |
Gene Name: | uncharacterized LOC100652999 |
Gene Aliases: |
- |
Location: |
Chr.12:53513984-53517608 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100652999 | NR_046221.1 | 3 | 833 | |
| AK027179.1 | 3 | 796 |
More Information
Set Membership: |
|

Genomic Map