Assay Details
Target Gene Details
Entrez Gene ID: | 92370 |
Gene Name: | 2-phosphoxylose phosphatase 1 |
Gene Aliases: |
ACPL2, HEL124, XYLP |
Location: |
Chr.3:141231825-141294903 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PXYLP1 | NM_001037172.2 | NP_001032249.1 | ||
| NM_001282728.1 | NP_001269657.1 | |||
| NM_152282.4 | NP_689495.1 | |||
| XM_017007511.1 | XP_016863000.1 | |||
| AB827640.1 | ||||
| AK074331.1 | BAB85053.1 | |||
| AK297790.1 | ||||
| AY358460.1 | AAQ88825.1 | |||
| BC035834.1 | AAH35834.1 | |||
| BX647621.1 | ||||
| CD101677.1 | ||||
| CX762355.1 | ||||
| DC409719.1 | ||||
| EU794677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591892 | Chr.3:141219704 - 141286732 on Build GRCh38 | Loss |
|
| nsv4039 | Chr.3:141230054 - 141263710 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map