Assay Details
Target Gene Details
Entrez Gene ID: | 1327 |
Gene Name: | cytochrome c oxidase subunit 4I1 |
Gene Aliases: |
COX IV-1, COX4, COX4-1, COXIV, COXIV-1 |
Location: |
Chr.16:85799567-85807003 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX4I1 | NM_001318786.1 | 5 | 769 | NP_001305715.1 |
| NM_001318788.1 | 5 | 726 | NP_001305717.1 | |
| NM_001318794.1 | 5 | 1030 | NP_001305723.1 | |
| NM_001318797.1 | 5 | 613 | NP_001305726.1 | |
| NM_001318802.1 | 5 | 708 | NP_001305731.1 | |
| NM_001861.4 | 5 | 748 | NP_001852.1 | |
| AK027136.1 | 1 | 3869 | ||
| BC008704.1 | 5 | 606 | AAH08704.1 | |
| BC021236.2 | 5 | 599 | AAH21236.1 | |
| BC047869.1 | 3 | 2194 | AAH47869.1 | |
| BC062437.1 | 5 | 627 | AAH62437.1 | |
| BE615615.1 | 4 | 567 | ||
| BE728651.1 | 5 | 610 | ||
| BP343428.1 | ||||
| CD677397.1 | 5 | 567 | ||
| M21575.1 | 5 | 595 | AAA99312.1 | |
| M34600.1 | 4 | 561 | AAA52059.1 | |
| U90915.1 | 5 | 631 | AAB51058.1 | |
| X54802.1 | 5 | 623 | CAA38573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833316 | Chr.16:85697332 - 85900033 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map