Assay Details
Target Gene Details
Entrez Gene ID: | 51388 |
Gene Name: | NIP7, nucleolar pre-rRNA processing protein |
Gene Aliases: |
CGI-37, HSPC031, KD93 |
Location: |
Chr.16:69339512-69343111 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIP7 | NM_001199434.1 | 4 | 956 | NP_001186363.1 |
| NM_016101.4 | 5 | 1097 | NP_057185.1 | |
| AB062398.1 | 5 | 799 | BAB93485.1 | |
| AB062479.1 | 5 | 799 | BAB93504.1 | |
| AB112439.2 | 5 | 937 | BAD05056.1 | |
| AF085360.1 | 5 | 812 | AAD40195.1 | |
| AF132971.1 | 5 | 812 | AAD27746.1 | |
| AF161528.1 | 4 | 670 | AAF29143.1 | |
| AK001158.1 | 4 | 648 | ||
| BC015941.1 | 5 | 824 | AAH15941.1 |
More Information
Set Membership: |
|

Genomic Map