Assay Details
Target Gene Details
Entrez Gene ID: | 767557 |
Gene Name: | SYS1-DBNDD2 readthrough (NMD candidate) |
Gene Aliases: |
C20orf169-DBNDD2 |
Location: |
Chr.20:45363169-45410610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYS1-DBNDD2 | NR_003189.2 | |||
| BI199175.1 | ||||
| DN994280.1 |
Target Gene Details
Entrez Gene ID: | 27296 |
Gene Name: | TP53 target 5 |
Gene Aliases: |
C20orf10, CLG01 |
Location: |
Chr.20:45373880-45378550 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53TG5 | NM_014477.2 | 3 | 291 | NP_055292.1 |
| XM_011528790.2 | 3 | 550 | XP_011527092.1 | |
| AB017802.1 | 3 | 374 | BAA76405.1 | |
| AK295308.1 | 3 | 312 | ||
| BC036785.1 | 3 | 290 | AAH36785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map