Assay Details
Target Gene Details
Entrez Gene ID: | 4818 |
Gene Name: | natural killer cell granule protein 7 |
Gene Aliases: |
GIG1, GMP-17, p15-TIA-1 |
Location: |
Chr.19:51371616-51372706 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKG7 | NM_005601.3 | NP_005592.1 | ||
| XM_005258955.3 | XP_005259012.1 | |||
| XM_006723228.3 | XP_006723291.1 | |||
| AI313472.1 | ||||
| BC015759.1 | AAH15759.1 | |||
| S69115.1 | AAB30078.1 |
More Information
Set Membership: |
|

Genomic Map