Assay Details
Target Gene Details
Entrez Gene ID: | 8533 |
Gene Name: | COP9 signalosome subunit 3 |
Gene Aliases: |
CSN3, SGN3 |
Location: |
Chr.17:17246624-17281303 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS3 | NM_001199125.1 | NP_001186054.1 | ||
| NM_001316354.1 | NP_001303283.1 | |||
| NM_001316355.1 | NP_001303284.1 | |||
| NM_001316356.1 | NP_001303285.1 | |||
| NM_001316357.1 | NP_001303286.1 | |||
| NM_001316358.1 | NP_001303287.1 | |||
| NM_003653.3 | NP_003644.2 | |||
| XM_005256837.4 | XP_005256894.1 | |||
| XM_005256840.3 | XP_005256897.1 | |||
| XM_005256842.4 | XP_005256899.1 | |||
| XM_017025246.1 | XP_016880735.1 | |||
| AF031647.1 | AAC14197.1 | |||
| AF098109.1 | AAD41247.1 | |||
| AK297706.1 | ||||
| AK302304.1 | ||||
| AK312476.1 | ||||
| AK316400.1 | ||||
| BC001891.1 | AAH01891.1 | |||
| BI520077.1 | ||||
| BQ018146.1 | ||||
| BU154357.1 | ||||
| BU171304.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833382 | Chr.17:17221015 - 17402369 on Build GRCh38 | Loss |
|
| nsv833380 | Chr.17:17185102 - 17337892 on Build GRCh38 | Loss |
|
| nsv469777 | Chr.17:17224071 - 17369384 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map