Assay Details
Target Gene Details
Entrez Gene ID: | 26122 |
Gene Name: | enhancer of polycomb homolog 2 |
Gene Aliases: |
EPC-LIKE |
Location: |
Chr.2:148644780-148788491 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPC2 | NM_015630.3 | 1 | 99 | NP_056445.3 |
| XM_011510941.2 | 1 | 310 | XP_011509243.1 | |
| AF286904.1 | 1 | 91 | AAK60500.1 | |
| AK125789.1 | 2 | 168 | ||
| BC093818.1 | AAH93818.1 | |||
| BC093820.1 | AAH93820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751831 | Chr.2:148393789 - 149249089 on Build GRCh38 | Gain |
|
| nsv508873 | Chr.2:148598370 - 148653458 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map