Assay Details
Target Gene Details
Entrez Gene ID: | 645121 |
Gene Name: | cyclin I family member 2 |
Gene Aliases: |
- |
Location: |
Chr.5:132747402-132754403 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNI2 | NM_001039780.3 | 2 | 532 | NP_001034869.1 |
| NM_001287252.1 | 2 | 532 | NP_001274181.1 | |
| NM_001287253.1 | 2 | 535 | NP_001274182.1 | |
| XM_017009725.1 | 2 | 578 | XP_016865214.1 | |
| AK131553.1 | 2 | 533 | BAD18687.1 | |
| BC132837.1 | 2 | 514 | ||
| BC136836.1 | 2 | 514 | ||
| BC144416.1 | 2 | 517 | ||
| BC144417.1 | 2 | 514 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv529021 | Chr.5:132682477 - 132750285 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map