Assay Details
Target Gene Details
Entrez Gene ID: | 4114 |
Gene Name: | MAGE family member B3 |
Gene Aliases: |
CT3.5 |
Location: |
Chr.X:30230436-30237493 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB3 | NM_002365.4 | 5 | 1807 | NP_002356.2 |
| XM_011545513.2 | 4 | 1414 | XP_011543815.1 | |
| XM_011545514.2 | 3 | 1381 | XP_011543816.1 | |
| AK057441.1 | 5 | 1807 | ||
| BC069784.1 | 1 | 1129 | AAH69784.1 | |
| BC074756.2 | AAH74756.1 | |||
| BC126320.1 | ||||
| BC126322.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs144072080] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map