Assay Details
Target Gene Details
Entrez Gene ID: | 10389 |
Gene Name: | sex comb on midleg-like 2 (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.X:18239313-18355089 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCML2 | NM_006089.2 | 10 | 1370 | NP_006080.1 |
| NR_033717.1 | 10 | 1370 | ||
| XM_006724458.1 | 10 | 1334 | XP_006724521.1 | |
| XM_017029219.1 | 10 | 1266 | XP_016884708.1 | |
| XM_017029220.1 | 10 | 1262 | XP_016884709.1 | |
| XM_017029221.1 | 10 | 1404 | XP_016884710.1 | |
| XM_017029222.1 | 4 | 648 | XP_016884711.1 | |
| AK299190.1 | 4 | 648 | ||
| AK303064.1 | 9 | 1236 | ||
| AK308140.1 | 4 | 644 | ||
| AK315116.1 | 10 | 1334 | ||
| BC051913.1 | 10 | 1312 | AAH51913.1 | |
| BC064617.1 | 10 | 1327 | AAH64617.1 | |
| Y18004.1 | 10 | 1304 | CAB38943.1 |
More Information
Set Membership: |
|

Genomic Map