Assay Details
Target Gene Details
Entrez Gene ID: | 6241 |
Gene Name: | ribonucleotide reductase regulatory subunit M2 |
Gene Aliases: |
R2, RR2, RR2M |
Location: |
Chr.2:10122568-10131419 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRM2 | NM_001034.3 | 10 | 1545 | NP_001025.1 |
| NM_001165931.1 | 10 | 1713 | NP_001159403.1 | |
| AK092671.1 | 10 | 1514 | ||
| AK123010.1 | 10 | 1481 | ||
| BC001886.1 | 10 | 1526 | AAH01886.1 | |
| BC028932.1 | 4 | 832 | AAH28932.1 | |
| BC030154.1 | 10 | 1531 | AAH30154.1 | |
| S40301.1 | 10 | 1676 | AAA09577.1 | |
| X59618.1 | 10 | 1676 | CAA42181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3715n100 | Chr.2:10014833 - 10135998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map