Assay Details
Target Gene Details
Entrez Gene ID: | 51605 |
Gene Name: | tRNA methyltransferase 6 |
Gene Aliases: |
CGI-09, GCD10, Gcd10p |
Location: |
Chr.20:5937832-5950558 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT6 | NM_001281467.1 | NP_001268396.1 | ||
| NM_015939.4 | NP_057023.2 | |||
| XM_017027875.1 | XP_016883364.1 | |||
| AB032979.1 | BAA86467.1 | |||
| AF132943.1 | AAD27718.1 | |||
| AK000613.1 | ||||
| AK300812.1 | ||||
| AK308648.1 | ||||
| AK309205.1 | ||||
| BC001262.1 | AAH01262.1 |
More Information
Set Membership: |
|

Genomic Map