Assay Details
Target Gene Details
Entrez Gene ID: | 3849 |
Gene Name: | keratin 2 |
Gene Aliases: |
CK-2e, K2e, KRT2A, KRT2E, KRTE |
Location: |
Chr.12:52644558-52652175 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT2 | NM_000423.2 | NP_000414.2 | ||
| BC096294.4 | AAH96294.1 | |||
| BC099643.1 | AAH99643.1 | |||
| BC099644.3 | AAH99644.1 | |||
| M99061.1 | AAC83410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428281 | Chr.12:52570654 - 52761443 on Build GRCh38 | Gain |
|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map