Assay Details
Target Gene Details
Entrez Gene ID: | 150468 |
Gene Name: | cytoskeleton associated protein 2 like |
Gene Aliases: |
- |
Location: |
Chr.2:112736349-112764677 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKAP2L | NM_001304361.1 | NP_001291290.1 | ||
| NM_152515.4 | NP_689728.3 | |||
| NR_130712.1 | ||||
| XM_011510666.2 | XP_011508968.1 | |||
| AK097948.1 | BAC05202.1 | |||
| AK292530.1 | ||||
| AK302875.1 | ||||
| AK316241.1 | ||||
| AL832036.1 | ||||
| BC036217.1 | AAH36217.1 | |||
| DA506407.1 |

Genomic Map