Assay Details
Target Gene Details
Entrez Gene ID: | 54878 |
Gene Name: | dipeptidyl peptidase 8 |
Gene Aliases: |
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141 |
Location: |
Chr.15:65442463-65517713 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833042 | Chr.15:65446079 - 65656395 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map