Assay Details
Target Gene Details
Entrez Gene ID: | 80279 |
Gene Name: | CDK5 regulatory subunit associated protein 3 |
Gene Aliases: |
C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553 |
Location: |
Chr.17:47967904-47981786 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK5RAP3 | NM_001278197.1 | NP_001265126.1 | ||
| NM_001278198.1 | NP_001265127.1 | |||
| NM_001278216.1 | NP_001265145.1 | |||
| NM_001278217.1 | NP_001265146.1 | |||
| NM_176096.2 | NP_788276.1 | |||
| XM_011525297.1 | XP_011523599.1 | |||
| XM_017025164.1 | XP_016880653.1 | |||
| XM_017025165.1 | XP_016880654.1 | |||
| XM_017025166.1 | XP_016880655.1 | |||
| AB062433.1 | BAB93496.1 | |||
| AF110322.1 | AAG39277.1 | |||
| AF217982.2 | AAG17225.1 | |||
| AF343090.1 | AAK69655.1 | |||
| AK000260.1 | ||||
| AK023722.1 | BAB14657.1 | |||
| AK223387.1 | BAD97107.1 | |||
| AK225274.1 | ||||
| AK300643.1 | ||||
| AK301739.1 | ||||
| BC009957.2 | AAH09957.1 | |||
| BC072435.1 | AAH72435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469808 | Chr.17:47972502 - 48151311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map