Assay Details
Target Gene Details
Entrez Gene ID: | 29889 |
Gene Name: | G protein nucleolar 2 |
Gene Aliases: |
HUMAUANTIG, NGP1, Ngp-1, Nog2, Nug2 |
Location: |
Chr.1:37566812-37595985 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNL2 | NM_001323623.1 | 14 | 2082 | NP_001310552.1 |
| NM_001323624.1 | 12 | 1693 | NP_001310553.1 | |
| NM_013285.2 | 13 | 1878 | NP_037417.1 | |
| AK130697.1 | 1 | 1890 | ||
| AK298358.1 | ||||
| AK312915.1 | 13 | 1832 | ||
| BC000107.2 | 13 | 1814 | AAH00107.1 | |
| BC009250.2 | 13 | 1814 | AAH09250.1 | |
| L05425.1 | 13 | 1794 | AAC37588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834746 | Chr.1:37537351 - 37701654 on Build GRCh38 | Loss |
|
| nsv524834 | Chr.1:37496012 - 37585984 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map