Assay Details
Target Gene Details
Entrez Gene ID: | 83473 |
Gene Name: | katanin catalytic subunit A1 like 2 |
Gene Aliases: |
- |
Location: |
Chr.18:46917602-47104795 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNAL2 | NM_031303.2 | NP_112593.2 | ||
| XM_005258357.4 | XP_005258414.1 | |||
| XM_005258358.4 | XP_005258415.1 | |||
| XM_006722554.3 | XP_006722617.1 | |||
| XM_011526219.2 | XP_011524521.1 | |||
| XM_011526220.1 | XP_011524522.1 | |||
| XM_011526221.2 | XP_011524523.1 | |||
| XM_011526223.2 | XP_011524525.1 | |||
| XM_017026028.1 | XP_016881517.1 | |||
| XM_017026029.1 | XP_016881518.1 | |||
| XM_017026030.1 | XP_016881519.1 | |||
| XM_017026032.1 | XP_016881521.1 | |||
| AK309134.1 | ||||
| BC034999.2 | AAH34999.2 |
Target Gene Details
Entrez Gene ID: | 51224 |
Gene Name: | transcription elongation factor B subunit 3B |
Gene Aliases: |
ELOA2, HsT832, TCEB3L |
Location: |
Chr.18:47032572-47035617 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB3B | NM_016427.2 | 1 | 1966 | NP_057511.2 |
| AB030834.1 | 1 | 1727 | BAA90665.1 | |
| AK314984.1 | 1 | 1946 | ||
| BC036022.1 | 1 | 1970 | AAH36022.1 | |
| BC103911.2 | 1 | 1614 | AAI03912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509693 | Chr.18:46942006 - 47047880 on Build GRCh38 | Insertion |
|
| nsv828225 | Chr.18:47032678 - 47033793 on Build GRCh38 | Gain |
|
| esv998188 | Chr.18:47032741 - 47033895 on Build GRCh38 | Gain |
|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| nsv833640 | Chr.18:47025905 - 47188501 on Build GRCh38 | Loss |
|
| nsv2284 | Chr.18:47024467 - 47043660 on Build GRCh38 | Insertion |
|
| nsv1056744 | Chr.18:46929089 - 47055381 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map