Assay Details
Target Gene Details
Entrez Gene ID: | 64399 |
Gene Name: | hedgehog interacting protein |
Gene Aliases: |
HIP |
Location: |
Chr.4:144645996-144742645 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHIP | NM_022475.2 | 2 | 1102 | NP_071920.1 |
| XM_005263178.4 | 2 | 971 | XP_005263235.1 | |
| XM_006714288.3 | 2 | 971 | XP_006714351.1 | |
| AK074711.1 | 2 | 718 | BAC11154.1 | |
| AK098525.1 | 2 | 720 | ||
| AK291849.1 | 2 | 442 | ||
| AY009317.1 | 2 | 942 | AAG35411.1 | |
| AY009951.1 | 2 | 1077 | AAG34731.1 | |
| AY358747.1 | 2 | 1052 | AAQ89107.1 | |
| BC009298.2 | 2 | 899 | AAH09298.1 | |
| BC025311.1 | 2 | 928 | AAH25311.1 |
More Information
Set Membership: |
|

Genomic Map