Assay Details
Target Gene Details
Entrez Gene ID: | 283651 |
Gene Name: | high mobility group nucleosomal binding domain 2 pseudogene 46 |
Gene Aliases: |
C15orf21, D-PCa-2 |
Location: |
Chr.15:45511136-45556730 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGN2P46 | NR_022014.1 | |||
| AK096745.1 | 5 | 617 | BAC04856.1 | |
| AY271964.1 | AAP12643.1 | |||
| AY271965.1 | AAP12644.1 | |||
| AY271966.1 | AAP12645.1 | |||
| AY271967.1 | AAP12646.1 |
Target Gene Details
Entrez Gene ID: | 7782 |
Gene Name: | solute carrier family 30 member 4 |
Gene Aliases: |
ZNT4, znT-4 |
Location: |
Chr.15:45479613-45523685 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A4 | NM_001321036.1 | NP_001307965.1 | ||
| NM_013309.5 | NP_037441.2 | |||
| XM_011521997.2 | XP_011520299.1 | |||
| XM_017022560.1 | XP_016878049.1 | |||
| XM_017022561.1 | XP_016878050.1 | |||
| XM_017022562.1 | XP_016878051.1 | |||
| AF025409.1 | AAB82561.1 | |||
| AK290874.1 | ||||
| BC026089.2 | AAH26089.1 | |||
| BM789500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892684 | Chr.15:45307172 - 46327637 on Build GRCh38 | Gain |
|
| esv3569553 | Chr.15:45366990 - 46080626 on Build GRCh38 | Gain |
|
| nsv1160292 | Chr.15:45351161 - 46175480 on Build GRCh38 | Duplication |
|
| nsv1045556 | Chr.15:45485198 - 45592656 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map