Assay Details
Target Gene Details
Entrez Gene ID: | 10606 |
Gene Name: | phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase |
Gene Aliases: |
ADE2, ADE2H1, AIRC, PAIS |
Location: |
Chr.4:56435749-56461368 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAICS | NM_001079524.1 | NP_001072992.1 | ||
| NM_001079525.1 | NP_001072993.1 | |||
| NM_006452.3 | NP_006443.1 | |||
| XM_011534366.1 | XP_011532668.1 | |||
| AK294821.1 | ||||
| BC010273.1 | AAH10273.1 | |||
| BC019255.1 | AAH19255.1 | |||
| BT006988.1 | AAP35634.1 | |||
| CF994644.1 | ||||
| CN337183.1 | ||||
| CR749824.1 | CAH18683.1 | |||
| D28431.1 | BAA05797.1 | |||
| DB503040.1 | ||||
| X53793.1 | CAA37801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161086 | Chr.4:56310907 - 56510457 on Build GRCh38 | Duplication |
|
| dgv5213n100 | Chr.4:56076751 - 56538345 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map