Assay Details
Target Gene Details
Entrez Gene ID: | 55716 |
Gene Name: | limb development membrane protein 1 like |
Gene Aliases: |
LIMR |
Location: |
Chr.12:49096551-49111535 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBR1L | NM_001300750.1 | NP_001287679.1 | ||
| NM_001300751.1 | NP_001287680.1 | |||
| NM_018113.3 | NP_060583.2 | |||
| XM_011538563.2 | 17 | 2479 | XP_011536865.1 | |
| XM_011538564.2 | 17 | 2852 | XP_011536866.1 | |
| XM_011538565.2 | 16 | 3101 | XP_011536867.1 | |
| XM_011538566.2 | 16 | 2075 | XP_011536868.1 | |
| XM_017019626.1 | 17 | 2727 | XP_016875115.1 | |
| XM_017019627.1 | 18 | 2456 | XP_016875116.1 | |
| XM_017019628.1 | 16 | 2152 | XP_016875117.1 | |
| XM_017019629.1 | 18 | 3311 | XP_016875118.1 | |
| XM_017019630.1 | 14 | 4287 | XP_016875119.1 | |
| XM_017019631.1 | 16 | 2015 | XP_016875120.1 | |
| AB033000.1 | BAA86488.1 | |||
| AF260728.1 | AAK60600.1 | |||
| AK001356.1 | BAA91646.1 | |||
| AK001651.1 | BAA91811.1 | |||
| AK093570.1 | ||||
| AL137599.1 | CAB70835.1 | |||
| AY358572.1 | AAQ88935.1 | |||
| BC008337.1 | AAH08337.1 | |||
| BC015015.1 | AAH15015.1 | |||
| BC031550.1 | AAH31550.1 |
More Information
Set Membership: |
|

Genomic Map