Assay Details
Target Gene Details
Entrez Gene ID: | 7401 |
Gene Name: | clarin 1 |
Gene Aliases: |
RP61, USH3, USH3A |
Location: |
Chr.3:150926163-150972999 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLRN1 | NM_001195794.1 | 4 | 2152 | NP_001182723.1 |
| NM_001256819.1 | 4 | 2285 | NP_001243748.1 | |
| NM_052995.2 | 4 | 794 | NP_443721.1 | |
| NM_174878.2 | 3 | 2113 | NP_777367.1 | |
| NR_046380.2 | 5 | 2303 | ||
| AF388366.1 | 4 | 794 | AAL09581.1 | |
| AF495717.1 | 3 | 1854 | AAM88774.1 |
More Information
Set Membership: |
|

Genomic Map