Assay Details
Target Gene Details
Entrez Gene ID: | 59082 |
Gene Name: | caspase recruitment domain family member 18 |
Gene Aliases: |
ICEBERG, UNQ5804, pseudo-ICE |
Location: |
Chr.11:105137721-105139734 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD18 | NM_021571.3 | NP_067546.1 | ||
| AI189838.1 | ||||
| AW295205.1 | ||||
| AY358231.1 | AAQ88598.1 | |||
| BC131610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052674 | Chr.11:104111970 - 107043401 on Build GRCh38 | Loss |
|
| esv2750978 | Chr.11:104094062 - 105475331 on Build GRCh38 | Gain |
|
| nsv468862 | Chr.11:104903287 - 105372644 on Build GRCh38 | Gain |
|
| nsv1038877 | Chr.11:104166130 - 105422422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map