Assay Details
Target Gene Details
Entrez Gene ID: | 54870 |
Gene Name: | glutamine rich 1 |
Gene Aliases: |
- |
Location: |
Chr.3:49029707-49094373 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRICH1 | NM_001320580.1 | 4 | 1597 | NP_001307509.1 |
| NM_001320581.1 | 4 | 1616 | NP_001307510.1 | |
| NM_001320582.1 | 3 | 1556 | NP_001307511.1 | |
| NM_001320583.1 | 3 | 1537 | NP_001307512.1 | |
| NM_001320584.1 | 3 | 1196 | NP_001307513.1 | |
| NM_001320585.1 | 3 | 1396 | NP_001307514.1 | |
| NM_017730.3 | 4 | 1681 | NP_060200.2 | |
| NM_198880.2 | 3 | 1621 | NP_942581.1 | |
| XM_011533863.1 | 3 | 1337 | XP_011532165.1 | |
| AK000266.1 | 1 | 82 | BAA91042.1 | |
| AK074313.1 | 4 | 1370 | BAB85047.1 | |
| AK093259.1 | 1 | 965 | ||
| AK292536.1 | 4 | 1434 | ||
| BC000978.2 | 2 | 1055 | AAH00978.2 | |
| BC098375.1 | 1 | 401 | AAH98375.1 | |
| BC110855.1 | 3 | 1299 | AAI10856.1 | |
| BC130340.1 | 3 | 1267 | ||
| BC130342.1 | 3 | 1267 |
More Information
Set Membership: |
|

Genomic Map