Assay Details
Target Gene Details
Entrez Gene ID: | 10826 |
Gene Name: | fatty acid hydroxylase domain containing 2 |
Gene Aliases: |
C5orf4 |
Location: |
Chr.5:154817018-154850685 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAXDC2 | NM_032385.4 | NP_115761.2 | ||
| XM_005268358.2 | XP_005268415.1 | |||
| XM_006714753.2 | XP_006714816.1 | |||
| XM_017008965.1 | XP_016864454.1 | |||
| XM_017008966.1 | XP_016864455.1 | |||
| AK023820.1 | ||||
| AK127944.1 | ||||
| AK295546.1 | ||||
| BC004506.2 | AAH04506.2 | |||
| BC007216.1 | AAH07216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830521 | Chr.5:154768007 - 154864706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map