Assay Details
Target Gene Details
Entrez Gene ID: | 400797 |
Gene Name: | long intergenic non-protein coding RNA 83 |
Gene Aliases: |
NCRNA00083 |
Location: |
Chr.1:178479514-178499623 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00083 | XM_017003086.1 | XP_016858575.1 | ||
| AK125993.1 | 4 | 612 | BAC86382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009797 | Chr.1:178489573 - 178512778 on Build GRCh38 | Gain |
|
| esv29324 | Chr.1:178479961 - 178512150 on Build GRCh38 | Gain |
|
| nsv517431 | Chr.1:178494456 - 178502359 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map