Assay Details
Target Gene Details
Entrez Gene ID: | 63931 |
Gene Name: | mitochondrial ribosomal protein S14 |
Gene Aliases: |
DJ262D12.2, HSMRPS14, MRP-S14, S14mt |
Location: |
Chr.1:175012958-175023455 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS14 | NM_022100.2 | 3 | 914 | NP_071383.1 |
| NR_037606.1 | 3 | 1006 | ||
| AK310915.1 | 3 | 976 | ||
| AL049705.1 | 3 | 884 | CAB41269.1 |
More Information
Set Membership: |
|

Genomic Map