Assay Details
Target Gene Details
Entrez Gene ID: | 219986 |
Gene Name: | olfactory receptor family 4 subfamily D member 11 |
Gene Aliases: |
OR4D11P |
Location: |
Chr.11:59503576-59504511 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D11 | NM_001004706.1 | 1 | 638 | NP_001004706.1 |
More Information
Set Membership: |
|

Genomic Map