Assay Details
Target Gene Details
Entrez Gene ID: | 25852 |
Gene Name: | armadillo repeat containing 8 |
Gene Aliases: |
GID5, HSPC056, S863-2, VID28 |
Location: |
Chr.3:138187248-138298389 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC8 | NM_001267041.2 | 21 | 3572 | NP_001253970.1 |
| NM_001267042.2 | 21 | 3464 | NP_001253971.1 | |
| NM_001282342.1 | 20 | 3495 | NP_001269271.1 | |
| NM_015396.5 | 23 | 3741 | NP_056211.2 | |
| XM_006713565.2 | 23 | 3701 | XP_006713628.1 | |
| XM_011512632.1 | 22 | 3608 | XP_011510934.1 | |
| XM_011512633.1 | 22 | 3662 | XP_011510935.1 | |
| XM_017006095.1 | 22 | 3462 | XP_016861584.1 | |
| XM_017006097.1 | 21 | 3408 | XP_016861586.1 | |
| XM_017006098.1 | 20 | 3354 | XP_016861587.1 | |
| XM_017006099.1 | 21 | 3445 | XP_016861588.1 | |
| AL096748.1 | 9 | 2762 | CAB46423.1 | |
| BQ719381.1 | 1 | 724 |
Target Gene Details
Entrez Gene ID: | 347736 |
Gene Name: | NME/NM23 family member 9 |
Gene Aliases: |
NM23-H9, TXL-2, TXL2, TXNDC6 |
Location: |
Chr.3:138261437-138331785 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME9 | NM_178130.3 | NP_835231.1 | ||
| XM_005247429.3 | 12 | 5441 | XP_005247486.1 | |
| XM_006713615.3 | 11 | 7323 | XP_006713678.1 | |
| XM_011512774.2 | 11 | 5327 | XP_011511076.1 | |
| XM_017006317.1 | 10 | 5376 | XP_016861806.1 | |
| XM_017006318.1 | 9 | 5704 | XP_016861807.1 | |
| AK094830.1 | BAC04430.1 | |||
| AY336746.1 | AAQ16116.1 | |||
| BC107054.2 | AAI07055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map