Assay Details
Target Gene Details
Entrez Gene ID: | 51634 |
Gene Name: | RNA binding motif protein, X-linked 2 |
Gene Aliases: |
CGI-79, Snu17 |
Location: |
Chr.X:130401969-130413343 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMX2 | NM_016024.2 | NP_057108.2 | ||
| AF078865.1 | AAD44497.1 | |||
| AF151837.1 | AAD34074.1 | |||
| AK292855.1 | ||||
| AL050405.1 | CAB43745.1 | |||
| BC003676.1 | ||||
| BC033750.1 | AAH33750.1 | |||
| BC105582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map