Assay Details
Target Gene Details
Entrez Gene ID: | 403 |
Gene Name: | ADP ribosylation factor like GTPase 3 |
Gene Aliases: |
ARFL3 |
Location: |
Chr.10:102673727-102714433 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL3 | NM_004311.3 | 6 | 2951 | NP_004302.1 |
| XM_017016260.1 | 6 | 3137 | XP_016871749.1 | |
| AF038193.1 | 1 | 650 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831969 | Chr.10:102651251 - 102812041 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map