Assay Details
Target Gene Details
Entrez Gene ID: | 126695 |
Gene Name: | keratinocyte differentiation factor 1 |
Gene Aliases: |
C1orf172 |
Location: |
Chr.1:26949556-26960484 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDF1 | NM_152365.2 | NP_689578.2 | ||
| XM_005245735.2 | XP_005245792.1 | |||
| XM_011540622.1 | XP_011538924.1 | |||
| AK091952.1 | BAC03775.1 | |||
| BC033143.1 | AAH33143.1 | |||
| CR749558.1 | CAH18355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508689 | Chr.1:26912462 - 26959506 on Build GRCh38 | Deletion |
|
| nsv1000012 | Chr.1:26714969 - 27141528 on Build GRCh38 | Gain |
|
| nsv834524 | Chr.1:26830184 - 26994502 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map