Assay Details
Target Gene Details
Entrez Gene ID: | 81624 |
Gene Name: | diaphanous related formin 3 |
Gene Aliases: |
AN, AUNA1, DIA2, DRF3, NSDAN, diap3, mDia2 |
Location: |
Chr.13:59665583-60163985 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH3 | NM_001042517.1 | NP_001035982.1 | ||
| NM_001258366.1 | NP_001245295.1 | |||
| NM_001258367.1 | NP_001245296.1 | |||
| NM_001258368.1 | NP_001245297.1 | |||
| NM_001258369.1 | NP_001245298.1 | |||
| XM_011535258.2 | XP_011533560.1 | |||
| XM_011535263.2 | XP_011533565.1 | |||
| XM_011535265.2 | XP_011533567.1 | |||
| XM_017020789.1 | XP_016876278.1 | |||
| AB244756.1 | ||||
| AB244757.1 | ||||
| AB244758.1 | ||||
| AK092024.1 | BAC03793.1 | |||
| AK298509.1 | ||||
| AK316474.1 | ||||
| AY750055.1 | AAW73254.1 | |||
| AY818645.1 | AAW78862.1 | |||
| BX649186.1 | CAE46204.1 |
Target Gene Details
Entrez Gene ID: | 100874195 |
Gene Name: | DIAPH3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:60012718-60044357 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH3-AS1 | NR_046539.2 | |||
| NR_051993.2 | ||||
| NR_051994.2 | ||||
| NR_109838.1 | ||||
| AI685327.1 | ||||
| AW468687.1 | ||||
| BX093461.1 | ||||
| BX398653.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561861 | Chr.13:59852754 - 60110259 on Build GRCh38 | Loss |
|
| nsv832622 | Chr.13:60024743 - 60223418 on Build GRCh38 | Gain+Loss |
|
| nsv561862 | Chr.13:60009783 - 60049569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map