Assay Details
Target Gene Details
Entrez Gene ID: | 1404 |
Gene Name: | hyaluronan and proteoglycan link protein 1 |
Gene Aliases: |
CRT1, CRTL1 |
Location: |
Chr.5:83638198-83721210 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAPLN1 | NM_001884.3 | 4 | 1023 | NP_001875.1 |
| XM_011543168.2 | 4 | 957 | XP_011541470.1 | |
| XM_017009051.1 | 5 | 1206 | XP_016864540.1 | |
| XM_017009052.1 | 4 | 819 | XP_016864541.1 | |
| XM_017009053.1 | 4 | 1003 | XP_016864542.1 | |
| XM_017009054.1 | 4 | 958 | XP_016864543.1 | |
| AK313713.1 | 4 | 978 | ||
| BC057808.1 | 3 | 779 | AAH57808.1 | |
| U43328.1 | 4 | 825 | AAA85216.1 | |
| X17405.1 | 4 | 1023 | CAA35462.1 | |
| X78076.1 | 4 | 1023 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4904 | Chr.5:83636117 - 83668059 on Build GRCh38 | Insertion |
|
| nsv830367 | Chr.5:83531833 - 83742562 on Build GRCh38 | Gain+Loss |
|
| nsv1015852 | Chr.5:83404850 - 84499122 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map