Assay Details
Target Gene Details
Entrez Gene ID: | 8645 |
Gene Name: | potassium two pore domain channel subfamily K member 5 |
Gene Aliases: |
K2p5.1, KCNK5b, TASK-2, TASK2 |
Location: |
Chr.6:39188971-39229475 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK5 | NM_003740.3 | 5 | 3170 | NP_003731.1 |
| XM_005249456.1 | 5 | 2894 | XP_005249513.1 | |
| XM_006715235.2 | 3 | 2433 | XP_006715298.1 | |
| AF084830.1 | AAC79458.1 | |||
| AK001897.1 | 1 | 1767 | ||
| BC060793.1 | 5 | 3170 | AAH60793.1 | |
| BU608155.1 | 1 | 631 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950167 | Chr.6:39188925 - 39197324 on Build GRCh38 | Deletion |
|
| nsv830644 | Chr.6:39148754 - 39358204 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map