Assay Details
Target Gene Details
Entrez Gene ID: | 6142 |
Gene Name: | ribosomal protein L18a |
Gene Aliases: |
L18A |
Location: |
Chr.19:17859878-17863324 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL18A | NM_000980.3 | NP_000971.1 | ||
| AK222647.1 | BAD96367.1 | |||
| AK297323.1 | ||||
| AK297358.1 | ||||
| AK311774.1 | ||||
| HY050156.1 | ||||
| L05093.1 | AAC18781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv978783 | Chr.19:17861250 - 17862337 on Build GRCh38 | Duplication |
|
| nsv833772 | Chr.19:17854364 - 18028528 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map