Assay Details
Target Gene Details
Entrez Gene ID: | 91689 |
Gene Name: | single-pass membrane protein with aspartate rich tail 1 |
Gene Aliases: |
C22orf32, DDDD, EMRE, dJ186O1.1 |
Location: |
Chr.22:42079687-42084284 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMDT1 | NM_033318.4 | NP_201575.3 | ||
| XM_011530509.2 | XP_011528811.1 | |||
| AK095636.1 | 1 | 473 | ||
| AL449243.1 | CAC15000.1 | |||
| BC024237.2 | AAH24237.1 | |||
| BI754670.1 | ||||
| CR456453.1 | CAG30339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508737 | Chr.22:42052007 - 42134336 on Build GRCh38 | Insertion |
|
| nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map