Assay Details
Target Gene Details
Entrez Gene ID: | 5420 |
Gene Name: | podocalyxin like |
Gene Aliases: |
Gp200, PC, PCLP, PCLP-1 |
Location: |
Chr.7:131500262-131556617 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PODXL | NM_001018111.2 | 9 | 2425 | NP_001018121.1 |
| NM_005397.3 | 8 | 2329 | NP_005388.2 | |
| BX641124.1 | 2 | 3161 | ||
| U97519.1 | 8 | 2327 | AAB61574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831134 | Chr.7:131455230 - 131614283 on Build GRCh38 | Gain |
|
| dgv6617n100 | Chr.7:131331684 - 131555085 on Build GRCh38 | Gain |
|
| esv7269 | Chr.7:131503589 - 131503787 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map