Assay Details
Target Gene Details
Entrez Gene ID: | 100528062 |
Gene Name: | ARMCX5-GPRASP2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:102599348-102717733 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMCX5-GPRASP2 | NM_001199818.1 | 7 | 1647 | NP_001186747.1 |
Target Gene Details
Entrez Gene ID: | 114928 |
Gene Name: | G protein-coupled receptor associated sorting protein 2 |
Gene Aliases: |
GASP2 |
Location: |
Chr.X:102712176-102717733 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRASP2 | NM_001004051.3 | 5 | 1515 | NP_001004051.1 |
| NM_001184874.2 | 5 | 1643 | NP_001171803.1 | |
| NM_001184875.2 | 4 | 1555 | NP_001171804.1 | |
| NM_001184876.2 | 5 | 1554 | NP_001171805.1 | |
| NM_138437.5 | 4 | 1427 | NP_612446.1 | |
| AK096067.1 | 1 | 765 | ||
| AK123832.1 | 4 | 1427 | ||
| AL832187.1 | 1 | 155 | ||
| BC013576.1 | 1 | 759 | AAH13576.1 | |
| BC036772.1 | 4 | 1191 | AAH36772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526716 | Chr.X:102582805 - 102934382 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map