Assay Details
Target Gene Details
Entrez Gene ID: | 6124 |
Gene Name: | ribosomal protein L4 |
Gene Aliases: |
L4 |
Location: |
Chr.15:66499315-66504855 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL4 | NM_000968.3 | NP_000959.2 | ||
| AB208977.1 | BAD92214.1 | |||
| AK125772.1 | ||||
| AK130784.1 | ||||
| AK223057.1 | BAD96777.1 | |||
| AK291117.1 | ||||
| AK291859.1 | ||||
| BC001365.2 | AAH01365.1 | |||
| BC009888.2 | AAH09888.1 | |||
| BC010151.2 | AAH10151.1 | |||
| BC014653.1 | AAH14653.1 | |||
| BC066925.1 | AAH66925.1 | |||
| BC095427.1 | AAH95427.1 | |||
| D23660.1 | BAA04887.1 | |||
| L20868.2 | AAA60281.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv974591 | Chr.15:66499279 - 66503530 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map