Assay Details
Target Gene Details
Entrez Gene ID: | 25973 |
Gene Name: | prolyl-tRNA synthetase 2, mitochondrial (putative) |
Gene Aliases: |
MT-PRORS, proRS |
Location: |
Chr.1:54756898-54764553 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARS2 | NM_152268.3 | 2 | 1811 | NP_689481.2 |
| AK025585.1 | 1 | 1493 | BAB15178.1 | |
| AL117473.1 | 1 | 1492 | CAB55948.1 | |
| BC007956.2 | 1 | 1726 | AAH07956.2 | |
| BC011758.2 | 2 | 1772 | AAH11758.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004778 | Chr.1:54635180 - 54764639 on Build GRCh38 | Gain |
|
| dgv166n100 | Chr.1:54741809 - 54804866 on Build GRCh38 | Gain |
|
| nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain |
|
| nsv546266 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv461594 | Chr.1:54593622 - 54757517 on Build GRCh38 | Gain |
|
| dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74946945] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map