Assay Details
Target Gene Details
Entrez Gene ID: | 275 |
Gene Name: | aminomethyltransferase |
Gene Aliases: |
GCE, GCST, GCVT, NKH |
Location: |
Chr.3:49416778-49422678 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMT | NM_000481.3 | NP_000472.2 | ||
| NM_001164710.1 | NP_001158182.1 | |||
| NM_001164711.1 | NP_001158183.1 | |||
| NM_001164712.1 | NP_001158184.1 | |||
| NR_028435.1 | ||||
| AK091738.1 | ||||
| AK096062.1 | ||||
| AK290600.1 | ||||
| AK293481.1 | ||||
| AK293594.1 | ||||
| AK294592.1 | ||||
| AK296177.1 | ||||
| AK301072.1 | 4 | 919 | ||
| BC007546.1 | AAH07546.2 | |||
| BC044792.1 | 1 | 1389 | AAH44792.1 | |
| D13811.1 | BAA02967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834687 | Chr.3:49411199 - 49594355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map