Assay Details
Target Gene Details
Entrez Gene ID: | 347733 |
Gene Name: | tubulin beta 2B class IIb |
Gene Aliases: |
PMGYSA, bA506K6.1 |
Location: |
Chr.6:3224261-3227734 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB2B | NM_178012.4 | 1 | 192 | NP_821080.1 |
| AK289433.1 | 1 | 113 | ||
| AK297568.1 | 1 | 111 | ||
| AK308806.1 | 1 | 110 | ||
| BC001352.2 | 1 | 192 | AAH01352.1 | |
| BC063610.1 | 1 | 95 | AAH63610.1 | |
| BT019930.1 | AAV38733.1 | |||
| CR456788.1 | CAG33069.1 |
More Information
Set Membership: |
|

Genomic Map