Assay Details
Target Gene Details
Entrez Gene ID: | 757 |
Gene Name: | transmembrane protein 50B |
Gene Aliases: |
C21orf4, HCVP7TP3 |
Location: |
Chr.21:33432486-33480009 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM50B | NM_006134.6 | NP_006125.2 | ||
| NR_040016.1 | ||||
| XM_011529746.1 | XP_011528048.1 | |||
| AF045606.2 | AAC05974.2 | |||
| AF086280.1 | ||||
| AK309977.1 | ||||
| AK310701.1 | ||||
| AK311870.1 | ||||
| AY358634.1 | AAQ88997.1 | |||
| AY820138.1 | AAV68502.1 | |||
| BC000569.2 | AAH00569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587396 | Chr.21:33449065 - 33462420 on Build GRCh38 | Loss |
|
| nsv528264 | Chr.21:33437700 - 33465479 on Build GRCh38 | Gain |
|
| esv3893406 | Chr.21:33437386 - 33486199 on Build GRCh38 | Loss |
|
| nsv3497 | Chr.21:33449048 - 33488305 on Build GRCh38 | Insertion |
|
| nsv834087 | Chr.21:33325890 - 33469002 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map